missing translation for 'onlineSavingsMsg'
Learn More

CD45 Rat anti-Mouse, PerCP, Clone: 30-F11, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 561047

 View more versions of this product

Product Code. 15850719

  • ONLINE EXCLUSIVE PRICE
    € 118.00
    € 117.40 / Each
    Save € 0.60
    0.51% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 30-F11 clone has been reported to react with all isoforms and both alloantigens of CD45, which is found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family, where the intracellular carboxy-terminal region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated as A, B, and C, respectively). CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD45
Monoclonal
0.2mg/mL
Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4
Rat
Affinity Purified
RUO
Mouse
IgG2b κ
Flow Cytometry
30-F11
PerCP
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Thymus / Spleen
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.