Learn More
CD45 Rat, PE-Cy7, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB40-PE7-100ug
 View more versions of this product
Product Code. 15981968
  • € 262.50 / 100µg
Specifications
CD45
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2b κ
Flow Cytometry
30-F11
PE-Cyanine7
NP_001255215
Rat
100 μg
Primary
Store at 2-4°C.