Learn More
CD45 Rat, PE-Cy7, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB40-PE7-25ug
This item is not returnable.
View return policy
Specifications
| CD45 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
| Ptprc | |
| Protein A/G Purified | |
| RUO | |
| 19264 | |
| IgG2b κ |
| Flow Cytometry | |
| 30-F11 | |
| PE-Cyanine7 | |
| NP_001255215 | |
| Rat | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |