missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R (B220) Rat anti-Mouse, BV750, Clone: RA3-6B2, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 747469
This item is not returnable.
View return policy
Description
CD45R (B220) Monoclonal antibody specifically detects CD45R (B220) in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD45R (B220) | |
| Monoclonal | |
| Brilliant Violet 750 | |
| B220; Ly-5; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C | |
| Mouse Abelson Leukemia Virus-Induced pre-B tumor cells | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| RA3-6B2 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction