missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Antibody (OX-33), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65000IR
This item is not returnable.
View return policy
Description
CD45RA Monoclonal specifically detects CD45RA in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD45RA | |
| Monoclonal | |
| DyLight 755 | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65000 is directed against a high molecular weight band of the leucocyte common antigen (LCA). The MRC OX-33 only labels B-cells among thoracic duct lymphocytes with little labelling in bone marrow and none on thymocytes (2). | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| OX-33 | |
| Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| PTPRC | |
| Purified rat spleen leucocyte common antigen | |
| 0.25 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction