missing translation for 'onlineSavingsMsg'
Learn More

CD45RA Mouse anti-Human, FITC, Clone: HI100, BD

Mouse Monoclonal Antibody

€ 79.18 - € 220.63

Specifications

Antigen CD45RA
Clone HI100
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15871429
View Documents
BD Biosciences
561882
25 Tests
ONLINE EXCLUSIVE PRICE
€ 79.20
€ 79.18 / Each
Save € 0.02
0.03% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15888208
View Documents
BD Biosciences
555488
100 Tests
ONLINE EXCLUSIVE PRICE
€ 221.00
€ 220.63 / Each
Save € 0.37
0.17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD45RA
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
HI100
Monoclonal
Mouse
Human
IgG2b κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.