Learn More
CD45RA Mouse anti-Human, PE, Clone: HI100, BD
Mouse Monoclonal Antibody
€ 79.18 - € 325.42
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | HI100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881429
|
BD Biosciences
561883 |
25 Tests |
€ 79.20 € 79.18 / Each Save € 0.02 0.03% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15898208
|
BD Biosciences
555489 |
100 Tests |
€ 344.00 € 325.42 / Each Save € 18.58 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.
Specifications
| CD45RA | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HI100 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2b κ | |
| Affinity Purified |