Learn More
CD45RA Mouse, anti-Human, Unconjugated, Clone: HI100, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65108-1-IG-500UG
Description
CD45, also known as protein tyrosine phosphatase, receptor type C, is a type I transmembrane protein expressed on the surface of all haematopoietic cells with the exception of erythrocytes and platelets (PMID: 3489673; 28615666). CD45 is a pan-haematopoietic cell marker and has been shown to be essential for T- and B-cell activation and signalling (PMID: 9429890; 16378097). CD45 exists as multiple isoforms due to alternative splicing of three exons (4, 5, and 6, designated A, B, and C) in the extracellular domain (PMID: 12414720). CD45RA is expressed on naïve T cells, B cells, and monocytes (PMID: 1830500; 14687231).
Specifications
| CD45RA | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| HI100 | |
| Unconjugated | |
| 100 μg | |
| 5788 | |
| Liquid |