missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Mouse anti-Human, BB700, Clone: MT4 (6B6), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746103
This item is not returnable.
View return policy
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Flow Cytometry.
Specifications
| CD45RB | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| MT4 (6B6) | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction