Learn More
CD45RO Mouse, anti-Human, Alexa Fluor 488, Clone: UCHL1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60097AD
This item is not returnable.
View return policy
Description
- The UCHL1 antibody reacts with an extracellular epitope on CD45RO, which is the shortest isoform of CD45, a type I transmembrane glycoprotein and member of the protein tyrosine phosphatase family (receptor class 1/6 subfamily)
- Alternative splicing of exons 4, 5 and 6 that encode the extracellular RA, RB and RC polypeptides of CD45 gives rise to up to 8 isoforms with molecular masses of 180 - 240 kDa
- Excision of all three exons generates the ∼180 kDa CD45RO isoform, which is expressed on activated and memory (but not naive) T cells, some B cell subsets, activated monocytes and macrophages, and granulocytes
- CD45RO enhances both T cell receptor- and B cell receptor-mediated activation and is a known ligand for CD22 on B cells
- The UCHL1 antibody has been used to identify T cell lymphomas and leukemia and is commonly used in combination with antibodies against CD45RA to discern memory and naive T cells
- The UCHL1 epitope is destroyed by treatment with neuraminidase or O-glycosidase
Specifications
| CD45RO | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 100 Tests | |
| Marmoset, Chimpanzee, Human | |
| Leukocytes, T Cells, T Cells, CD4+, T Cells, CD8+ |
| Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry | |
| UCHL1 | |
| Mouse | |
| Primary | |
| IgG2a κ |