Learn More
CD45RO Mouse anti-Human , BV480, Clone: UCHL1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566143
This item is not returnable.
View return policy
Description
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry
Specifications
| CD45RO | |
| Monoclonal | |
| Brilliant Violet 480 | |
| CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200 | |
| Human IL-2-dependent T-cell line | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| UCHL1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| The antibody was conjugated with BD Horizon BV480 under optimum conditions, and unconjugated antibody and free BD Horizon BV480 were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |