Learn More
CD46 Antibody (7D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26679-50ul
 View more versions of this product
Product Code. 30043831
  • € 343.00 / 50µL
Description
CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD46
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
7D12
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
A synthesized peptide derived from Human CD46 [UniProt P15529]
50 μL
Cancer, Cell Biology, Cellular Markers, Immunology
4179
Store at -20 to -70C. Avoid freeze-thaw cycles.