missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Antibody (M177), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51131
This item is not returnable.
View return policy
Description
CD46 Monoclonal specifically detects CD46 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunoassay.
Specifications
| CD46 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional, Immunoassay | |
| CD46 | |
| Purified MCP | |
| 0.05 mg | |
| Cancer, Cell Biology, Cellular Markers, Immunology | |
| 4179 | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, Immunoassay | |
| M177 | |
| Unconjugated | |
| AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction