missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Antibody (MEM-258), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-599
This item is not returnable.
View return policy
Description
CD46 Monoclonal specifically detects CD46 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications
| CD46 | |
| Monoclonal | |
| FITC | |
| AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen | |
| Mouse | |
| 66 kDa | |
| 100 Tests | |
| Cancer, Cell Biology, Cellular Markers, Immunology | |
| 4179 | |
| Human, Bovine | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| MEM-258 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
| CD46 | |
| HPB-ALL human T cell line | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-258 recognizes an epitope on SCR4 (the membrane-proximal SCR) domain of CD46 (Membrane cofactor protein). CD46 is 56-66 kDa dimeric transmembrane protein expressed on T and B lymphocytes, platelets, monocytes, granulocytes, endothelial cells, epithelial cells and fibroblast; it is negative on erythrocytes. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction