missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (974214), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1866H
This item is not returnable.
View return policy
Description
CD47 Monoclonal antibody specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CD47 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 961 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 974214 | |
| Flow Cytometry, CyTOF-ready | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Mouse myeloma cell line, NS0-derived mouse CD47 protein, Gln19-Pro158, Accession # NP_034711 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction