missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (B6H12.2), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31106PECY55
This item is not returnable.
View return policy
Description
CD47 Monoclonal specifically detects CD47 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD47 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CD47 | |
| This antibody was raised against CD47. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| B6H12.2 | |
| Flow Cytometry | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Mouse | |
| Protein G purified | |
| Immunology, Signal Transduction | |
| 961.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction