missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (CD47/6362R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08218AF700
This item is not returnable.
View return policy
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| CD47 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| A recombinant fragment (around aa 18-135) of human CD47 protein (exact sequence is proprietary) (Uniprot: Q08722) | |
| 100 μg | |
| Immunology, Signal Transduction | |
| 961 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| CD47/6362R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction