missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (IAP/964) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08974
This item is not returnable.
View return policy
Description
CD47 Monoclonal antibody specifically detects CD47 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| CD47 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Recombinant full-length human CD47 protein (Uniprot: Q08722) | |
| 0.1 mg | |
| Immunology, Signal Transduction | |
| 961 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| IAP/964 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction