Learn More
CD47 Monoclonal Antibody (B6H12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804843S
This item is not returnable.
View return policy
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow CytometrySpecifications
| CD47 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09237 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4316 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry | |
| B6H12 | |
| Unconjugated | |
| MMP7 | |
| 9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
| Human recombinant protein fragment corresponding to amino acids 95-267 of human MMP7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |