Learn More
CD47 Mouse anti-Human, BUV737, Clone: B6H12, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 749177
Product Code. 17240735
  • € 389.00 / 50µg
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD47
Monoclonal
Brilliant Ultraviolet 737
IAP; CDw149; gp42; MER6; Neurophilin; OA3 ; Rh-related antigen
Purified human placental RGD-binding proteins
RUO
Human
IgG1 κ
Flow Cytometry
B6H12
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody.