missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48/SLAMF2 Antibody (CD48/8360R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21026PCP
This item is not returnable.
View return policy
Description
CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD48/SLAMF2 | |
| Monoclonal | |
| PerCP | |
| BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| Recombinant fragment (around aa 59-178) of human CD48/SLAMF2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
| 962 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CD48/8360R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction