missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48/SLAMF2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76556DL594
This item is not returnable.
View return policy
Description
CD48/SLAMF2 Polyclonal antibody specifically detects CD48/SLAMF2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| CD48/SLAMF2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 594 | |
| BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human SLAMF2. The immunogen is located within amino acids 90 - 140 of SLAMF2. Amino Acid Squence: SKVQKEDNSTYIMRVLKKT | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
| 962 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction