Learn More
CD49c Mouse anti-Human, BUV395, Clone: C3 II.1, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744522
Description
The C3 II.1 monoclonal antibody specifically recognizes CD49c which is also known as Integrin alpha 3 (α3 integrin). CD49c is encoded by ITGA3 /(Integrin subunit alpha 3). CD49c isa 150 kDa type I transmembrane glycoprotein which non-covalently associates with integrin β1 (CD29) to form the α3β1 (CD49c/CD29, VLA-3) complex. CD49c is expressed mostly on endothelial and epithelial cells (basal epidermal layers). It is not expressedon platelets and is expressed weakly on peripheral blood leucocytes, including T cells, B cells, and monocytes. The CD49c/CD29 complex serves as an adhesive receptor for kalinin or epiligrin. This interaction may be important for thymocyte interaction with thymic epithelium.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD49c | |
| C3 II.1 | |
| Brilliant Ultraviolet 395 | |
| ITGA3, CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.