Learn More
CD49d Mouse anti-Baboon, Equine, Feline, Human, Brilliant Ultraviolet 737, Clone: 9F10, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612850
Description
The 9F10 monoclonal antibody specifically reacts with the integrin α 4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β 1 (CD29) or β 7. The α 4β 1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The α β 7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α 4 chain-containing integrins. This clone cross-reacts with a subset of peripheral blood lymphocytes, monocytes, and some granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on leukocytes is similar to that observed with human peripheral blood leukocytes.
Specifications
| CD49d | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 9F10 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Baboon, Equine, Feline, Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.