missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49e Armenian Hamster anti-Mouse, BUV395, Clone: HM alpha 5-1, BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 743115
This item is not returnable.
View return policy
Description
CD49e Monoclonal antibody specifically detects CD49e in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD49e | |
| Monoclonal | |
| 0.2 mg/mL | |
| Armenian Hamster | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| HM alpha 5-1 | |
| Brilliant Ultraviolet 395 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction