missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49e Armenian Hamster anti-Mouse, Oligo, Clone: HM alpha 5-1, BD™ AbSeq ™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 940421
This item is not returnable.
View return policy
Description
CD49e Monoclonal antibody specifically detects CD49e in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD49e | |
| Monoclonal | |
| Oligo | |
| ITGA5; Integrin α5 chain; VLA-5α-chain; VLA5a VLA-5a; Fnra | |
| Affinity-purified mouse VLA-5 protein (α5β1, CD49e/CD29) | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
| Single Cell 3' Sequencing | |
| Clone HM alpha 5-1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction