missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (CL12420), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21215-25ul
This item is not returnable.
View return policy
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CD5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL12420 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:1000- 1:2500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:1000- 1:2500 | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| This antibody was developed against a Synthetic Peptide of P06127. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
| 921 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction