Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | CD5 |
| Applications | Western Blot, Immunohistochemistry |
| Gene Alias | CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 |
| Immunogen | Thymocytes followed by Sezary T cells. |
| Primary or Secondary | Primary |
| Form | Purified |