missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASS1 Monoclonal Antibody (OTI8C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809122S
This item is not returnable.
View return policy
Description
ASS1 Monoclonal antibody specifically detects ASS1 in Human samples. It is validated for Western BlotSpecifications
| ASS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16455 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4255 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8C4 | |
| Unconjugated | |
| MGMT | |
| AA408052; arginino succinate synthetase; argininosuccinate synthase; argininosuccinate synthase 1; argininosuccinate synthetase; argininosuccinate synthetase 1; arginosuccinate synthetase 1; ASS; Ass1; Ass-1; ASSA; citrulline-aspartate ligase; citrulline--aspartate ligase; citrullinemia; CN; CTLN1; fold; mutant arginino succinate synthetase; RP11-618A20.2 | |
| Full length human recombinant protein of human MGMT produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction