Learn More
ESRRA Monoclonal Antibody (OTI2G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809112S
Encompass_Preferred
 View more versions of this product
Product Code. 17551014
  • € 242.00 / 30µL
Description
ESRRA Monoclonal antibody specifically detects ESRRA in Human samples. It is validated for Western Blot
Specifications
Specifications
ESRRA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P23771
Mouse
Affinity chromatography
RUO
2625
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2G5
Unconjugated
GATA3
ERR alpha; ERR1; ERRa; ERRalpha; ERR-alpha; Errra; ESRL1; ESRRA; ESRRA1; ESRRA2; estrogen receptor related 1; estrogen receptor-like 1; estrogen related receptor alpha; estrogen related receptor alpha 1; estrogen related receptor, alpha; estrogen-related nuclear receptor alpha; estrogen-related receptor alpha; estrogen-related receptor, alpha; Estrra; HERR1; LOW QUALITY PROTEIN: steroid hormone receptor ERR1; NR3B1; Nuclear receptor subfamily 3 group B member 1; orphan nuclear receptor; steroid hormone receptor ERR1; Steroid hormone receptor ERR1 (Estrogen-related receptor, alpha) (ERR-alpha) (Estrogen receptor-like 1)
Human recombinant protein fragment corresponding to amino acids 155-443 of human GATA3 produced in E.coli.