Learn More
CD5 Mouse anti-Human, Clone: OTI6D6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802401
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter′s Syndrome.Specifications
| CD5 | |
| Monoclonal | |
| Unconjugated | |
| CD5 | |
| LEU1, T1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 921 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry | |
| OTI6D6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P06127 | |
| CD5 | |
| Full length human recombit protein of human CD5 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.