missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD50 / ICAM-3 Mouse, Clone: 186-2G9, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3385-MSM1-P1
This item is not returnable.
View return policy
Specifications
| CD50 / ICAM-3 | |
| Cell surface | |
| 186-2G9 | |
| Unconjugated | |
| NP_002153 | |
| Mouse | |
| 110-160kDa | |
| Protein A/G Purified | |
| RUO | |
| 3385 | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| ICAM3 | |
| Stimulated human leukocytes | |
| HL-60 or THP-1 cells. Lymph node, tonsil | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction