Learn More
CD50 Mouse anti-Human, Brilliant Violet 605, Clone: TU41, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742305
Description
The TU41 monoclonal antibody specifically binds to the intracellular adhesion molecule-3 (ICAM-3) which is also known as CD50. ICAM-3 is a 110-120 kDa type I membrane glycoprotein expressed on all leukocytes (platelets and erythrocytes are ICAM-3-negative). ICAM-3 is also expressed on epidermal Langerhans cells and endothelial cells. It predominantly interacts with LFA-1 (CD11a/CD18) without requiring activation as the initial dendritic cell-T lymphocyte interaction. ICAM-3 is an important LFA-1 ligand that functions in the initiation of the immune/inflammatory response and also plays a role in chemotaxis.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
| CD50 | |
| TU41 | |
| Brilliant Violet 605 | |
| ICAM-3, ICAM3, Intercellular adhesion molecule 3 | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.