Learn More
CD52 Antibody (Campath-1H), PE - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52645PE
Product Code. 18656906
  • € 668.00 / 0.10mL
Description
CD52 Monoclonal antibody specifically detects CD52 in Human, Cynomolgus Monkey, Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)In vivo Assay, CyTOF, Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD52
Monoclonal
PE
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Human lymphocytes.
0.1 mL
Cancer, Immunology
1043
Store at 4°C in the dark.
IgG κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Campath-1H
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Cynomolgus Monkey, Macaque