missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD52 Antibody (HI186) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-311
This item is not returnable.
View return policy
Description
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| CD52 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 2 ug/ml, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, CyTOF-ready | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| Mouse | |
| 28 kDa | |
| 0.1 mg | |
| Cancer, Immunology | |
| 1043 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| HI186 | |
| Unconjugated | |
| P31358 | |
| CD52 | |
| Human tonisl (NM_001803.2). | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody HI186 reacts with CD52 (CAMPATH-1), a 21-28 kDa glycoprotein containing a large N-linked carbohydrate moiety; mature CD52 molecule is actually much smaller (approx. 8-9 kDa). CD52 is expressed at high levels on lymphocytes, monocytes/macrophages and in male reproductive tract. HLDA VI; WS Code BP 523 HLDA VI; WS Code T 6T-057 | |
| Store at 4C. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction