missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD52 Antibody (YTH 34.5-G2b (Campath-1G)), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52704AF647
This item is not returnable.
View return policy
Description
CD52 Monoclonal antibody specifically detects CD52 in Human, Cynomolgus Monkey, Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)In vivo Assay, CyTOF
Specifications
| CD52 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| Human peripheral mononuclear cells enriched for T-cells. | |
| 0.1 mL | |
| Cancer, Immunology | |
| 1043 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| YTH 34.5-G2b (Campath-1G) | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction