missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD52 Monoclonal Antibody (11/305/44), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528412
This item is not returnable.
View return policy
Description
Mouse anti Pig CD52, clone 11/305/44 is a monoclonal antibody recognizing the porcine homologue of human CD52, a approximately 19 kDa antigen expressed by mature lymphocytes, monocytes and dendritic cells.
CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.
Specifications
| CD52 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| 0 | |
| CD52 | |
| Porcine thymocytes. | |
| 100 μg | |
| Primary | |
| Pig | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 11/305/44 | |
| FITC | |
| CD52 | |
| AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100873164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion