missing translation for 'onlineSavingsMsg'
Learn More

CD53 Antibody (161-2), Biotin, Novus Biologicals™

Product Code. p-200057475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
100 μg
Unit Size:
0.1mL
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613056 100 μg 100µL
18634276 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613056 Supplier Novus Biologicals Supplier No. NBP247859B100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD53 Monoclonal antibody specifically detects CD53 in Human, Baboon (Negative), Equine (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD53
Applications Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 161-2
Conjugate Biotin
Formulation PBS
Gene Alias antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53
Host Species Mouse
Immunogen Stimulated human leukocytes
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 963
Target Species Human, Baboon (Negative), Equine (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.