missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Antibody (63-5A3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11477MFV500
This item is not returnable.
View return policy
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow Cytometry
Specifications
| CD53 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 63-5A3 | |
| Flow Cytometry | |
| antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
| Human Sezary cells | |
| 0.1 mL | |
| Immunology | |
| 963 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction