Learn More
CD53 Antibody (MEM-53), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB500-393H
Product Code. 18515441
  • ONLINE EXCLUSIVE PRICE
    € 635.00
    € 600.60 / 0.10mL
    Save € 34.40
    5% Off
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD53
Monoclonal
HRP
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53
Leukocytes of patient suffering from a LGL-type leukemia.
0.1 mL
Immunology
963
Store at 4°C in the dark.
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
MEM-53