Learn More
CD53 Mouse anti-Human, Brilliant Violet 510, Clone: HI29, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742963
Description
The HI29 monoclonal antibody specifically recognizes CD53. CD53 is a 35-42 kDa type III transmembrane glycoprotein that is expressed on all leucocytes, including plasma cells, but not on platelets, erythrocytes, and non-hematopoietic tissues. CD53 mediates signal transduction in monocytes and B cells. It has the broadest reactivity of all pan-leukocyte reagents used in immunohistology applications.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
| CD53 | |
| HI29 | |
| Brilliant Violet 510 | |
| tetraspanin-25, TSPAN25, tspan-25, MOX44 | |
| Human Peripheral Blood Mononuclear Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.