missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551130
This item is not returnable.
View return policy
Description
CD53 / TSPAN-25 Polyclonal Antibody detects endogenous levels of CD53 / TSPAN-25 protein.
CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
Specifications
| CD53 | |
| Polyclonal | |
| Unconjugated | |
| Cd53 | |
| AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12508, 24251, 963 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P19397, P24485, Q61451 | |
| Cd53 | |
| Synthesized peptide derived from the Internal region of human CD53. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction