missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD54 / ICAM-1 Mouse, APC, Clone: 15.2, enQuireBio™
Mouse Monoclonal Antibody
€ 196.35 - € 459.90
Specifications
| Antigen | CD54 / ICAM-1 |
|---|---|
| Clone | 15.2 |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15912098
|
EnquireBio
QAB47-APC-25Tests |
25 Tests |
€ 196.35
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15902098
|
EnquireBio
QAB47-APC-100Tests |
100 Tests |
€ 459.90
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD54 / ICAM-1 | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 3383 | |
| IgG1 | |
| Protein A/G Purified |
| 15.2 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_000192 | |
| ICAM1 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title