Learn More
CD55/DAF Antibody (278810), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB20091APCCY7
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry
Specifications
| CD55/DAF | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 278810 | |
| Flow Cytometry | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| NS0-derived recombinant human CD55/DAF, Asp35-Ser353, Accession # P08174.4 | |
| 0.1 mL | |
| Immunology | |
| 1604 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |