missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD55/DAF Antibody (583905), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5376PEATT594
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD55/DAF | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 583905 | |
| Flow Cytometry | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD55/DAF, Met1-Pro359, Accession # Q61475 | |
| 0.1 mL | |
| Immunology | |
| 1604 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction