missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD55/DAF Antibody (67) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 380.00
Specifications
| Antigen | CD55/DAF |
|---|---|
| Clone | 67 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677906
|
Novus Biologicals
NBP2-50226-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637256
|
Novus Biologicals
NBP2-50226 |
0.1 mL |
€ 380.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
| CD55/DAF | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 1604 | |
| IgG1 | |
| Protein A purified |
| 67 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| Fibronectin purified human monocytes . | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title