missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD55/DAF Antibody (F4-29D9), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47963DL594
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
Specifications
| CD55/DAF | |
| Monoclonal | |
| DyLight 594 | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| Human umbilical vein endothelial cells (HUVEC) | |
| 0.1 mL | |
| Immunology | |
| 1604 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| F4-29D9 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction