Learn More
Invitrogen™ CD55 Monoclonal Antibody (67)

Mouse Monoclonal Antibody
€ 522.00 - € 654.00
Specifications
| Antigen | CD55 |
|---|---|
| Clone | 67 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11520103
![]() |
Invitrogen™
MA182066 |
100 μg |
€ 522.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11590153
|
Invitrogen™
MA182424 |
200 μg |
€ 654.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. This antibody is not recommend for staining erythrocytes in flow cytometry applications. A suggested positive control for immunohistochemical applications is human tonsil. Mouse anti Human CD55 antibody, clone 67 recognizes the human CD55 cell surface antigen, a GPI linked molecule also known as decay accelerating factor (DAF). Mouse anti Human CD55 antibody, clone 67 recognizes the human CD55 cell surface antigen, a GPI linked molecule also known as decay accelerating factor (DAF).
CD55 (decay-accelerating factor, DAF) is a GPI-anchored membrane glycoprotein that protects autologous cells from classical and alternative pathway of complement cascade. Bidirectional interactions between CD55 and CD97 are involved in T cell regulation and CD55 can regulate complement when bound to CD97. In tumors, CD55 promotes neoangiogenesis, tumorigenesis, invasiveness and evasion of apoptosis. CD55 is a 70 kDa glycoprotein (in erythrocytes) anchored in the membrane by glycosylphosphatidylinositol tail. In other cells, the apparent molecular weight is somewhat larger for CD55 and has a substantial content of O-glycans. CD55 binds to activated C4b or C3b complement fragments on the cell surface, preventing the assembly and accelerating the decay of both classical and alternative pathways. CD55 carries the Cromer related blood group antigens. Two alternatively spliced transcripts encoding CD55 have been identified, and the predominant transcript encodes a membrane-bound protein expressed on cells exposed to plasma component proteins but an alternatively spliced transcript produces a soluble protein present at much lower levels. CD55 has been studied to have a role in prostate cancer growth and survival.Specifications
| CD55 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P08174 | |
| 1604 | |
| K562 cells. | |
| Antibody |
| 67 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| CD55 | |
| CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC | |
| CD55 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.