missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Mouse anti-Human, Oligo, Clone: NCAM16.2 (also known as NCAM 16), BD™ AbSeq ™
Mouse Monoclonal Antibody
Brand: BD Biosciences 940007
This item is not returnable.
View return policy
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD56 | |
| Monoclonal | |
| Oligo | |
| NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 | |
| Immunoaffinity-enriched adult human brain NCAM | |
| 25 Tests | |
| Primary | |
| Human | |
| IgG2b κ |
| Single Cell 3' Sequencing | |
| Clone NCAM16.2 (also known as NCAM 16) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 7099 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction