missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (NK1/7565), Novus Biologicals™
Mouse Monoclonal Antibody
€ 275.00 - € 530.00
Specifications
| Antigen | CD57 |
|---|---|
| Clone | NK1/7565 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682105
|
Novus Biologicals
NBP3-20275-100ug |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668964
|
Novus Biologicals
NBP3-20275-20ug |
20 μg |
€ 275.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD57 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Immunology | |
| 10mM PBS with 0.05% BSA | |
| 27087 | |
| IgG2 λ | |
| Protein A purified |
| NK1/7565 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title